Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
PCR Plate, 96-well, semi-skirted, flat deck, black lettering, red
Thermo Scientific™

PCR Plate, 96-well, semi-skirted, flat deck, black lettering, red

The Thermo Scientific 96-Well, Red with Black Lettering, Semi-Skirted Flat Deck PCR Plates are compatible with most thermal cyclers andRead more
Have Questions?
Catalog NumberQuantity
AB1400RL25 plates
Catalog number AB1400RL
Price (USD)
168.65
Online exclusive
194.00
Save 25.35
Each
Quantity:
25 plates
Request bulk or custom format
Price (USD)
168.65
Online exclusive
194.00
Save 25.35
Each
The Thermo Scientific 96-Well, Red with Black Lettering, Semi-Skirted Flat Deck PCR Plates are compatible with most thermal cyclers and ABI platforms, including sequencers, with no adapters necessary. The Thermo Scientific PCR plastics are designed, manufactured, and tested to ensure PCR performance. The plates have 0.3 mL maximum well volume when used with adhesive and heat seals.

Features of Thermo Scientific 96-Well Semi-Skirted Flat Deck PCR Plates

• Directly compatible with all major major thermal cyclers, including ABI platforms and sequencers with no adapters necessary
• Red color and black lettering facilitate sample tracking
• Flat plate deck facilitates sealing and handling
• Available as SuperPlate providing 4X more rigidity for superior robotic handling
• Raised rim design around each well enables secure sealing and safeguards against evaporation
• Uniform, thin well walls deliver maximum and consistent heat transfer
• Black lettered versions feature black alpha-numeric lettering to allow for easy well identification
• White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
• Cut corner: A12

Applications


• PCR

Related Products
PCR Plate, 96-well, semi-skirted, flat deck
PCR Plate, 96-well, semi-skirted, flat deck, white
PCR Plate, 96-well, semi-skirted, flat deck, blue
PCR Plate, 96-well, semi-skirted, flat deck, green
PCR Plate, 96-well, semi-skirted, flat deck, black
PCR Plate, 96-well, semi-skirted, flat deck, purple
PCR Plate, 96-well, semi-skirted, flat deck, red
PCR Plate, 96-well, semi-skirted, flat deck, yellow
PCR Plate, 96-well, semi-skirted, flat deck, black lettering, white
PCR Plate, 96-well, semi-skirted, flat deck, black lettering, blue
PCR Plate, 96-well, semi-skirted, flat deck, black lettering, green
PCR Plate, 96-well, semi-skirted, flat deck, black lettering, purple
PCR Plate, 96-well, semi-skirted, flat deck, black lettering, yellow
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 plates
Reaction SpeedStandard
Skirt StyleSemi
Volume (Metric) Working0.2 mL
BarcodeNo Barcode
ColorRed
MaterialPolypropylene
No. of Wells96
Unit SizeEach