Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
PCR Plate, 96-well, semi-skirted, flat deck, white
Thermo Scientific™

PCR Plate, 96-well, semi-skirted, flat deck, white

The Thermo Scientific 96-Well, White, Semi-Skirted Flat Deck PCR Plates are compatible with most thermal cyclers and ABI platforms, includingRead more
Have Questions?
Catalog NumberQuantity
AB1400W25 plates
Catalog number AB1400W
Price (USD)
170.65
Online exclusive
201.00
Save 30.35
Each
Quantity:
25 plates
Request bulk or custom format
Price (USD)
170.65
Online exclusive
201.00
Save 30.35
Each
The Thermo Scientific 96-Well, White, Semi-Skirted Flat Deck PCR Plates are compatible with most thermal cyclers and ABI platforms, including sequencers, with no adapters necessary. White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection. The Thermo Scientific PCR plastics are designed, manufactured, and tested to ensure PCR performance. The plates have 0.3 mL maximum well volume when used with adhesive and heat seals.

Features of Thermo Scientific 96-Well Semi-Skirted Flat Deck PCR Plates

• Directly compatible with all major major thermal cyclers, including ABI platforms and sequencers with no adapters necessary
• White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
• Flat plate deck facilitates sealing and handling
• Available as SuperPlate providing 4X more rigidity for superior robotic handling
• Raised rim design around each well enables secure sealing and safeguards against evaporation
• Uniform, thin well walls deliver maximum and consistent heat transfer
• Black lettered versions feature black alpha-numeric lettering to allow for easy well identification
• Cut corner: A12

Applications


• PCR
• qPCR/Real-Time PCR
• Sequencing

Related Products
PCR Plate, 96-well, semi-skirted, flat deck
PCR Plate, 96-well, semi-skirted, flat deck, blue
PCR Plate, 96-well, semi-skirted, flat deck, green
PCR Plate, 96-well, semi-skirted, flat deck, black
PCR Plate, 96-well, semi-skirted, flat deck, purple
PCR Plate, 96-well, semi-skirted, flat deck, red
PCR Plate, 96-well, semi-skirted, flat deck, yellow
PCR Plate, 96-well, semi-skirted, flat deck, black lettering, white
PCR Plate, 96-well, semi-skirted, flat deck, black lettering, blue
PCR Plate, 96-well, semi-skirted, flat deck, black lettering, green
PCR Plate, 96-well, semi-skirted, flat deck, black lettering, purple
PCR Plate, 96-well, semi-skirted, flat deck, black lettering, red
PCR Plate, 96-well, semi-skirted, flat deck, black lettering, yellow
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 plates
Reaction SpeedStandard
Skirt StyleSemi
Volume (Metric) Working0.2 mL
BarcodeNo Barcode
ColorWhite
MaterialPolypropylene
No. of Wells96
Unit SizeEach

Frequently asked questions (FAQs)

Why should I use Thermo Scientific white plastics in qPCR?

White plates and tubes reflect significantly more signal back to the qPCR instrument's detector than traditional clear plates. The improved signal reflection ensures that even the lowest levels of fluorescence are detected. White well walls also prevent signal from passing through to the cycler block where it can be inconsistently reflected or absorbed. This keeps variations in the cycler block from affecting qPCR data. Using Thermo Scientific white PCR plastics increases sensitivity and reduces variability within the qPCR assay.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.