Are Thermo Scientific adhesive seals free of DNase, RNase, and human DNA?
Whilst Thermo Scientific adhesive seals are manufactured in a clean facility, they are not certified free of DNase, RNase, and human DNA.