Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
SuperPlate PCR Plate, 96-well, semi-skirted, white
Thermo Scientific™

SuperPlate PCR Plate, 96-well, semi-skirted, white

Thermo Scientific 96-Well Semi-Skirted Flat Deck PCR Plates, White, are compatible with most thermal cyclers and ABI platforms, including sequencers,Read more
Have Questions?
Catalog NumberQuantity
AB2400W25 plates
Catalog number AB2400W
Price (USD)
218.65
Online exclusive
251.00
Save 32.35
Each
Quantity:
25 plates
Price (USD)
218.65
Online exclusive
251.00
Save 32.35
Each
Thermo Scientific 96-Well Semi-Skirted Flat Deck PCR Plates, White, are compatible with most thermal cyclers and ABI platforms, including sequencers, with no adapters necessary. Thermo Scientific PCR plastics are designed, manufactured, and tested to ensure PCR performance. These white plates have 0.3 mL maximum well volume when used with adhesive and heat seals.

Features of 96-Well Semi-Skirted Flat Deck PCR Plates:

• White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
• Directly compatible with all major major thermal cyclers, including ABI platforms and sequencers with no adapters necessary
• Flat plate deck facilitates sealing and handling
• SuperPlate provides 4X more rigidity for superior robotic handling
• Raised rim design around each well enables secure sealing and safeguards against evaporation
• Uniform, thin well walls deliver maximum and consistent heat transfer
• Cut corner: A12

Applications

• qPCR/Real-Time PCR
• Sequencing

Related Products
SuperPlate PCR Plate, 96-well, semi-skirted
SuperPlate PCR Plate, 96-well, semi-skirted, blue
SuperPlate PCR Plate, 96-well, semi-skirted, green
SuperPlate PCR Plate, 96-well, semi-skirted, purple
SuperPlate PCR Plate, 96-well, semi-skirted, red
SuperPlate PCR Plate, 96-well, semi-skirted, yellow
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 plates
Reaction SpeedStandard
Skirt StyleSemi
Volume (Metric) Well0.3 mL
Volume (Metric) Working0.2 mL
BarcodeNo Barcode
ColorWhite
MaterialPolypropylene
No. of Wells96
Product LineSuperPlate
Unit SizeEach

Frequently asked questions (FAQs)

Why should I use Thermo Scientific white plastics in qPCR?

White plates and tubes reflect significantly more signal back to the qPCR instrument's detector than traditional clear plates. The improved signal reflection ensures that even the lowest levels of fluorescence are detected. White well walls also prevent signal from passing through to the cycler block where it can be inconsistently reflected or absorbed. This keeps variations in the cycler block from affecting qPCR data. Using Thermo Scientific white PCR plastics increases sensitivity and reduces variability within the qPCR assay.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.