Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
SuperPlate PCR Plate, 96-well, low profile, skirted
Thermo Scientific™

SuperPlate PCR Plate, 96-well, low profile, skirted

The Thermo Scientific 96-Well Full-Skirted Plate, Robotic's signature full skirt surround provides an even surface for robotic arms to grip.Read more
Have Questions?
Catalog NumberQuantity
AB280025 plates
Catalog number AB2800
Price (USD)
176.65
Online exclusive
210.00
Save 33.35
Each
Quantity:
25 plates
Request bulk or custom format
Price (USD)
176.65
Online exclusive
210.00
Save 33.35
Each
The Thermo Scientific 96-Well Full-Skirted Plate, Robotic's signature full skirt surround provides an even surface for robotic arms to grip. The SuperPlate design is four times stronger than standard polypropylene plates due to an improved design and a specially selected polymer that minimize warping during thermal cycling and after heat sealing. The plates have thin walls to reduce evaporation and a cut corner at the H1 location. Use these 96-Well Full-Skirted Plates in PCR and qPCR applications.

Features of 96-Well Full-Skirted Plates, Robotic:

• ANSI:SBS footprint and stackable for use in automated systems
• SuperPlate provides 4X more rigidity for superior robotic handling
• Low profile to reduce dead space and increase PCR efficiency
• Raised rim design around each well enables secure sealing and safeguards against evaporation
• Uniform, thin well walls deliver maximum and consistent heat transfer for equally high performance from every sample
• 0.2 mL maximum well volume (when used with adhesive and heat seals)
• Cut Corner: H1

Applications

• PCR
• qPCR/Real-Time PCR

Related Products
PCR Plate, 96-well, robotic
PCR Plate, 96-well, robotic, white
SuperPlate PCR Plate, 96-well, low profile, skirted, white
SuperPlate PCR Plate, 96-well, low profile, skirted, blue
SuperPlate PCR Plate, 96-well, low profile, skirted, green
SuperPlate PCR Plate, 96-well, low profile, skirted, purple
SuperPlate PCR Plate, 96-well, low profile, skirted, red
SuperPlate PCR Plate, 96-well, low profile, skirted, yellow
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 plates
Reaction SpeedLow profile
Skirt StyleSkirted
Volume (Metric) Working0.1 mL
BarcodeNo Barcode
ColorClear
MaterialPolypropylene
No. of Wells96
Product LineSuperPlate
Unit SizeEach

Frequently asked questions (FAQs)

When should I use a Thermo Scientific low profile PCR plastic?

We recommend Thermo Scientific low profile PCR plates and tubes for reaction volumes below 20 µL. The low profile design minimizes the airspace between the PCR reaction and reduces evaporation effects, thereby increasing PCR efficiency in low volume reactions.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What is a low profile Thermo Scientific plate/tube?

Low profile Thermo Scientific PCR plastics have shorter wells than the standard profile plastics, decreasing the “dead space” between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Plate warping is causing problems with the robot in my automated processes. How can I solve this?

We recommend Thermo Scientific Armadillo PCR plates which are designed for robotic handling with high rigidity and resistance. Combining the rigidity of a polycarbonate frame with thin walled polypropylene wells, the Armadillo plates provide superior thermal cycling performance under all conditions without warping.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Are Thermo Scientific ABgene plastics endotoxin-free?

They are not certified endotoxin-free. However, their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Are Thermo Scientific ABgene plastics sterile?

Thermo Scientific Abgene plastics are not typically sterilized as their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA. In contrast, during typical non-clean room production, plastics are exposed to many contaminants including dust, bacterial cells, and DNA. The plastics are then sterilized to kill bacteria and inactivate RNases and DNases, but sterilization does not remove dust or DNA contamination. The dust particles left behind can inhibit PCR, and the damaged DNA fragments can still act as templates, leading to non-specific amplification.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.