Verso 1-step RT-qPCR Kit, with separate ROX vial, 400 reactions - FAQs
Verso 1-step RT-qPCR Kit, with separate ROX vial, 400 reactions - FAQs
View additional product information for Verso 1-step RT-qPCR Kit, with separate ROX vial - FAQs (AB4100A, AB4100C)
2 product FAQs found
What is the RT Enhancer that is included in all Verso kits?
The RT Enhancer is a unique enzyme that degrades contaminating genomic DNA at the start of the RT step. It is equally as effective as DNase I treatment but more convenient as it eliminates the need for a separate DNase I incubation prior to RT-PCR.
Do I need to treat my RNA with DNase I before running RT-qPCR with Verso cDNA Synthesis Kit?
No, you do not need to treat your RNA with DNase I before running RT-qPCR with the Verso cDNA Synthesis Kit. The RT enhancer included in the Verso kit degrades any contaminating double-stranded DNA during the reverse transcription process, eliminating the need for a separate DNase I treatment.