ABsolute Blue QPCR Mix, SYBR Green, with separate ROX vial
Thermo Scientific™

ABsolute Blue QPCR Mix, SYBR Green, with separate ROX vial

**********************Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
AB4166B
also known as AB-4166/B
1,600 Reactions
AB4167A400 Reactions
Catalog number AB4166B
also known as AB-4166/B
Price (USD)
3,485.00
Each
No. of Reactions:
1,600 Reactions
Price (USD)
3,485.00
Each
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of ABsolute Blue QPCR Mix, SYBR Green, and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific ABsolute Blue qPCR SYBR Green Mixes are optimized 2X master mixes for SYBR Green chemistry. The master mixes incorporate an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof. Absolute Blue qPCR Mixes include Thermo-Start Taq hot start enzyme for increased sensitivity.

Highlights

• High performance—ABsolute sensitivity, consistency & convenience
• Enhanced contrast and reduced errors—easily identify correctly aliquotted reaction mix in all plates and tubes
• Universal—compatible with all major qPCR platforms
• Adaptable—available with ROX or Fluorescein passive reference dyes

Applications

• Gene expression analysis
• siRNA validation
• Genotyping

Related Products
ABsolute Blue QPCR Mix, low ROX
ABsolute Blue QPCR Mix, with separate ROX vial
ABsolute Blue QPCR Mix, ROX
ABsolute Blue QPCR Mix, SYBR Green, low ROX
ABsolute Blue QPCR Mix, SYBR Green, ROX
ABsolute Blue QPCR Mix, SYBR Green, with separate ROX vial
ABsolute Blue QPCR Mix, SYBR Green, fluorescein
For Research Use Only. Not for use in diagnostic procedures.
Specifications
CompatibilityBlock-Based qPCR Instruments
For Use With (Equipment)BioRad CFX96™, MJ Chromo4, MJ Opticon, Eppendorf MasterCycler RealPlex, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, Corbett RotorGene
FormatTube
No. of Reactions1,600 Reactions
PolymeraseTaq DNA Polymerase
Product LineABsolute Blue
Product TypeReal Time PCR SYBR Master Mix
Quantity1,600 reactions
Sample TypeDNA, dsDNA
Sufficient For1600 Reactions of 25 μL Each
Volume16 x 1.25 mL
Concentration2X
Detection MethodSYBR
For Use With (Application)Gene Expression, Genotyping, siRNA Validation
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Contains:
• 16 x 1.25 mL ABsolute Blue QPCR SYBR Green Mix (2X)
• 100 μL MgCl2 (1M)
• 25 μL ROX Reference Dye (1 mM)
Sufficient for 1600 x 25 μL reactions.

Store at -20°C