Animal-Free Murine Dendritic Cell Cytokine Package, PeproTech®
Gibco™
Animal-Free Murine Dendritic Cell Cytokine Package, PeproTech®
The Animal-Free Murine Dendritic Cell Cytokine Package contains the key components required for the culturing of mouse dendritic cells. TheRead more
Have Questions?
Catalog NumberQuantity
AF-MDC1 pack
Catalog number AF-MDC
Price (USD)
1,126.00
Each
Quantity:
1 pack
Price (USD)
1,126.00
Each
The Animal-Free Murine Dendritic Cell Cytokine Package contains the key components required for the culturing of mouse dendritic cells.
The package contains the following two vials:
• 100 μg Animal-Free Murine IL-4 (Cat. No. AF-214-14-100UG)
• 100 μg Animal-Free Murine GM-CSF (Cat. No. AF-315-03-100UG)
IL-4 is a pleiotropic cytokine that regulates diverse T- and B-cell responses including cell proliferation, survival, and gene expression. Produced by mast cells, T cells, and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells.
GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells.
Specifications
For Use With (Application)Functional Assay
FormulationLyophilized
Quantity1 pack
SpeciesMurine
Product TypeDendritic Cell Cytokine Package
Unit SizeEach