What magnetic stand should I purchase for manual extraction? Cat. No. AM10027 or Cat. No. AM10050?
It is a personal preference as to which magnetic stand to purchase. Cat. No. AM10027 can be easier if using a pipettor for liquid handling, as beads clump to the side, Cat. No. AM10050 if using a robotic liquid handler, as beads clump in a ring.
Which U-bottom microplates are compatible with Magnetic Stand-96 (Cat. No. AM10027)?
Most standard 96-well microplates with U-bottom, including deep well format, should be compatible with Magnetic Stand-96. The following KingFisher 96-well plates have been validated to be compatible with Magnetic Stand-96:
- Cat. No. 95040455
- Cat. No. 95040460