RNAqueous™ Spin Cartridges - FAQs
RNAqueous™ Spin Cartridges - FAQs
View additional product information for RNAqueous™ Total RNA Isolation Kit - FAQs (AM10051G, AM1912)
1 product FAQs found
Can the Filter Cartridges within the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908) be ordered separately?
Yes, we do offer the Spin Cartridges (Cat. No. AM10051G) as a stand-alone product, and they would be compatible with the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908).