Thin-walled, frosted lid, RNase-free PCR tubes (0.2 mL) - FAQs
Thin-walled, frosted lid, RNase-free PCR tubes (0.2 mL) - FAQs
View additional product information for Thin-walled, frosted lid, RNase-free PCR tubes (0.2 mL) - FAQs (AM12225)
3 product FAQs found
How should I store Thin-walled, frosted lid, RNase-free PCR tubes (0.2 mL) (Cat. No. AM12225)?
We recommend storing Thin-walled, frosted lid, RNase-free PCR tubes (0.2 mL) (Cat. No. AM12225) at room temperature.
How many tubes are supplied in Thin-walled, frosted lid, RNase-free PCR tubes (0.2 mL) (Cat. No. AM12225)?
Thin-walled, frosted lid, RNase-free PCR tubes (0.2 mL) (Cat. No. AM12225) includes 1000 tubes. Each tube can hold 0.2 mL volume.
In Thin-walled, frosted lid, RNase-free PCR tubes (0.2 mL) (Cat. No. AM12225), why does the cap have a frosted surface?
The frosted cap provides a suitable surface for detailed labeling of the tube.