PCR Tubes & Caps, RNase-free, 0.2 mL (8-strip format) - FAQs
PCR Tubes & Caps, RNase-free, 0.2 mL (8-strip format) - FAQs
View additional product information for PCR Tubes & Caps, RNase-free, 0.2 mL (8-strip format) - FAQs (AM12230)
1 product FAQs found
Do the PCR Tubes & Caps, RNase-free, 0.2 mL, (8-strip format) (Cat. No. AM12230) have domed caps?
Yes, the PCR Tubes & Caps, RNase-free, 0.2 mL, (8-strip format) (Cat. No. AM12230) have domed caps.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.