RNase-Free Elution Tubes (2.0 mL) - FAQs
RNase-Free Elution Tubes (2.0 mL) - FAQs
View additional product information for RNase-Free Elution Tubes (2.0 mL) - FAQs (AM12480)
1 product FAQs found
Can the collection tubes within the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908) be ordered separately?
Yes, the Elution Tubes (2.0 mL) (Cat. No. AM12480) are the same that are used in the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908).