MAXIscript™ T7 Transcription Kit, 30 reactions - FAQs
MAXIscript™ T7 Transcription Kit, 30 reactions - FAQs
View additional product information for MAXIscript™ T7 Transcription Kit - FAQs (AM1312, AM1314M, AM1314)
2 product FAQs found
What probe kits do you offer for nonisotopic probe labeling and radiolabeled probe labeling?
We offer two nonisotopic probe labeling kits:
- The BrightStar Psoralen-Biotin Nonisotopic Labeling Kit (Cat. No. AM1480)
- MAXIscript Kits for in vitro transcription (Cat. Nos. AM1308 through AM1326)
We offer three radiolabeled probe labeling kits:
- MAXIscript Kits for in vitro transcription (Cat. Nos. AM1308 through AM1326)
- KinaseMax 5' End-Labeling Kit (Cat. No. AM1520)
- DECAprime II DNA Labeling Kit (Cat. No. AM1455)
What kits should I use to isolate RNA from my sample?
The best isolation kit to use will depend greatly on your application, but, in general, the kits below produce RNA input compatible with the Ion Total RNA-Seq Kit v2 workflow:
Whole Transcriptome (polyA-selected or rRNA-depleted RNA is highly recommended, but total RNA is acceptable)
- Poly A-selected RNA
• Dynabeads mRNA DIRECT Micro Kit (Cat. No. 61021)
• MicroPoly(A)Purist Kit (Cat. No. AM1919)
- rRNA-depleted RNA
• RiboMinus Eukaryote System v2 (Cat. No. A15026)
• Low Input RiboMinus Eukaryote System v2 (Cat. No. A15027)
- Total RNA
• TRI Reagent
• TRizol
• RecoverAll MagMax
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.