MAXIscript™ SP6/T7 Transcription Kit, 100 Reactions - FAQs
MAXIscript™ SP6/T7 Transcription Kit, 100 Reactions - FAQs
View additional product information for MAXIscript™ SP6/T7 Transcription Kit - FAQs (AM1322)
1 product FAQs found
What probe kits do you offer for nonisotopic probe labeling and radiolabeled probe labeling?
We offer two nonisotopic probe labeling kits:
- The BrightStar Psoralen-Biotin Nonisotopic Labeling Kit (Cat. No. AM1480)
- MAXIscript Kits for in vitro transcription (Cat. Nos. AM1308 through AM1326)
We offer three radiolabeled probe labeling kits:
- MAXIscript Kits for in vitro transcription (Cat. Nos. AM1308 through AM1326)
- KinaseMax 5' End-Labeling Kit (Cat. No. AM1520)
- DECAprime II DNA Labeling Kit (Cat. No. AM1455)