mMESSAGE mMACHINE™ T7 ULTRA Transcription Kit - FAQs
View additional product information for mMESSAGE mMACHINE™ T7 ULTRA Transcription Kit - FAQs (AM1345)
1 product FAQs found
Does the Lipofectamine MessengerMAX Transfection Reagent kit include reagents for in vitro transcription of mRNA?
Although it does not, we recommend the mMESSAGE mMACHINE T7 ULTRA Transcription Kit (Cat. No. AMB1345) to generate capped and tailed mRNA for highest mRNA stability.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.