MEGAshortscript™ T7 Transcription Kit - FAQs
MEGAshortscript™ T7 Transcription Kit - FAQs
View additional product information for MEGAshortscript™ T7 Transcription Kit - FAQs (AM1354)
2 product FAQs found
What size template should I use with the MEGAshortscript T7 kit?
The MEGAshortscript Kit is designed to function best with templates that code for RNA transcripts in the 20 to 500 nt range. The kit can be used to produce longer RNA but the Invitrogen MEGAscript Kit is more economical for this purpose.
I am planning on ordering Invitrogen GeneArt CRISPR T7 Strings DNA. What do you recommend I use to in vitro transcribe gRNA?
We recommend using our Invitrogen Megashortscript T7 Transcription Kit in order to produce ready-to-use in vitro transcribed gRNA.