Do you offer kits for probe construction for small RNA and miRNA analysis?
Yes, we offer our mirVana miRNA Probe Construction Kit (Cat. No. AM1550) and our mirVana Probe and Marker Kit (Cat. No. AM1554).
Search
View additional product information for mirVana™ miRNA Probe Construction Kit - FAQs (AM1550)
1 product FAQs found