mirVana™ PARIS™ RNA and Native Protein Purification Kit - FAQs
mirVana™ PARIS™ RNA and Native Protein Purification Kit - FAQs
View additional product information for mirVana™ PARIS™ RNA and Native Protein Purification Kit - FAQs (AM1556)
1 product FAQs found
I'd like to isolate RNA and protein from the same sample. Do you have a kit for this?
Yes, you can use TRIzol Reagent or the mirVana PARIS RNA and Native Protein Purification Kit.