I need a recommendation for a MagMAX kit for automated viral RNA isolation from Animal Health samples such as serum, nasal fluid, and swab samples. What would you recommend?
Cat. No. AM1836 is a good choice if you want to process 50 µL of samples. If you want to maximize the input amount and use more than that (up to 300 µL) we recommend using the MagMAX RNA/DNA Pathogen kit (Cat. No. 4462359). The buffers and reagents included in these MagMAX kits are identical, but the volumes are larger for the MagMAX Pathogen RNA/DNA kit allowing for more sample input.