RNAqueous™-96 Total RNA Isolation Kit - FAQs
RNAqueous™-96 Total RNA Isolation Kit - FAQs
View additional product information for RNAqueous™-96 Total RNA Isolation Kit - FAQs (AM1920)
2 product FAQs found
What are the differences in the RNAqueous kits you offer?
RNaqueous kits are based on a rapid, filter-based RNA isolation system that does not require phenol, chloroform, or other toxic organic chemicals. The RNAqueous Total RNA Isolation Kit is designed for sample sizes of 10-75 mg of tissue or 10^6 to 10^7 cells, whereas the RNAqueous-Midi Total RNA Isolation Kit is designed for tissue samples of up to 0.5 g or 10^9 cells. The RNAqueous-Micro Total RNA Isolation Kit is optimized for the purification of total RNA from micro-sized samples such as 10-500,000 cultured cells, as few as 10 microdissected cells, or up to 10 mg tissue. The RNAqueous-96 Total RNA Isolation Kit utilizes a 96-well plate format for high-throughput RNA isolation from 100 to 2 x 10^6 cells or 0.1 to 1.5 mg of tissue. The RNAqueous-4PCR Total RNA Isolation Kit is designed for RNA isolation from small tissue samples (0.5-75 mg) and small- to medium-sized cultured cell samples (approximately 100-10^7 cells).
Can you please recommend the best manifold to use with the RNAqueous-96 Total RNA Isolation Kit (Cat. No. AM1920)?
The EveryPrep Universal Vacuum Manifold (Cat. No. K211101) is compatible with the RNAqueous-96 Total RNA Isolation Kit.