Poly(A) Polymerase (cloned) 2 U/μL - FAQs
Poly(A) Polymerase (cloned) 2 U/μL - FAQs
View additional product information for Poly(A) Polymerase (cloned) 2 U/μL - FAQs (AM2030)
1 product FAQs found
Is the Poly(A) Polymerase (cloned) (Cat. No. AM2030) from E. coli?
PolyA polymerase (cloned) is purified from an E. coli strain overexpressing E. coli Poly(A) Polymerase I. Here is the link to the product information sheet: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/4393885_PlyAPlymrseCln_PI.pdf