BSA, acetylated (20 mg/mL)
Invitrogen™
BSA, acetylated (20 mg/mL)
Ambion® Acetylated BSA is used as a carrier protein and as an enzyme stabilizer in which the absence of contaminantsRead more
Have Questions?
Catalog NumberQuantity
AM26146 x 25 mg
Catalog number AM2614
Price (USD)
351.00
Each
Quantity:
6 x 25 mg
Price (USD)
351.00
Each
Ambion® Acetylated BSA is used as a carrier protein and as an enzyme stabilizer in which the absence of contaminants such as nucleases and proteases is essential. Six tubes of 25 mg are provided, at a concentration of 20 mg/mL. The acetylation process inactivates any trace nuclease activity that might be present. While the acetylation process does change BSA's binding characteristics and makes it inhibitory in PCR reactions, it is routinely added to restriction digests. The molecular weight is ∼66 kD. Acetylated BSA is rigorously tested for contaminating nonspecific endonuclease, exonuclease, protease, and RNase activity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
PackagingTube
Recommended StorageStore at –20°C.
Concentration20 mg/mL
Physical FormLiquid
Product LineAmbion
Quantity6 x 25 mg
Unit SizeEach