Oligo (dT) Primer (50 μM)
Invitrogen™

Oligo (dT) Primer (50 μM)

These are the same Ambion® primers that are currently included in the RETROscript® Kit (SKU# AM1710). They are provided atRead more
Have Questions?
Catalog NumberQuantity
AM5730G80 μL
Catalog number AM5730G
Price (USD)
179.65
Online exclusive
191.00
Save 11.35
Each
Quantity:
80 μL
Price (USD)
179.65
Online exclusive
191.00
Save 11.35
Each
These are the same Ambion® primers that are currently included in the RETROscript® Kit (SKU# AM1710). They are provided at a stock concentration of 50 μM and are functionally tested using the RETROscript® Kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Product TypePrimer
Quantity80 μL
Shipping ConditionDry Ice
Concentration50 μM
PrimerRandom
Product LineAmbion
Unit SizeEach
Contents & Storage
Store at –20°C.

Frequently asked questions (FAQs)

How does the Anchored Oligo(dT)20 Primer differ from standard oligo(dT) primers?

Anchored Oligo(dT) primers have 2 random bases at the 3' end of the stretch of Ts. The first random base is either an A, G, or C, while the second can be any of the 4 standard nucleotides, i.e., A, T, G or C. The use of anchored oligo(dT) primers results in increased cDNA synthesis yields.