Product Image
Invitrogen™

pMIR-REPORT™ miRNA Expression Reporter Vector System

The Ambion™ pMIR-REPORT™ miRNA Expression Reporter Vector System is for the measurement of miRNA expression in cells. It contains twoRead more
Have Questions?
Catalog NumberQuantity
AM57951 kit
Catalog number AM5795
Price (USD)
652.00
Each
Quantity:
1 kit
Price (USD)
652.00
Each
The Ambion™ pMIR-REPORT™ miRNA Expression Reporter Vector System is for the measurement of miRNA expression in cells. It contains two mammalian expression vectors—one for cloning miRNA targets and evaluating miRNA regulation, and the other for the normalization of transfection efficiency.

• Clone miRNA targets and evaluate miRNA regulation
• Screen putative miRNA target sequences
• Includes both reporter and control vectors

The pMIR-REPORT™ miRNA Expression Reporter Vector System provides accurate, quantitative, in-cell measurement of miRNA expression. This validated reporter system contains two mammalian expression vectors. The pMIR-REPORT™ Luciferase miRNA Expression Reporter Vector contains firefly luciferase under the control of a mammalian promoter/terminator system, with an miRNA target cloning region downstream of the luciferase translation sequence. This vector is optimized for cloning miRNA targets and evaluating miRNA regulation. A second vector, pMIR-REPORT™ Beta-galactosidase Reporter Control Vector, is provided for normalizing transfection efficiency.

Applications:
pMIR-REPORT™ Luciferase is designed for cloning and testing putative miRNA binding sites. pMIR-REPORT™ Luciferase can be transfected into mammalian cells to evaluate endogenous miRNA expression, or used to evaluate the up- or down-regulation resulting from the transfection of Pre-miR™ miRNA Molecules or Anti-miR™ miRNA Inhibitor Molecules, respectively.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cloning MethodRestriction Enzyme ⁄ MCS
Constitutive or Inducible SystemConstitutive
Delivery TypeTransfection
GeneBeta-Gal (lacZ), Luciferase
Product LineAmbion, pMIR-REPORT
Product TypeRNAi Expression Vector Kit
Quantity1 kit
RNAi TypemiRNA
Selection Agent (Eukaryotic)Puromycin
Shipping ConditionDry Ice
VectorpMIR
PromoterCMV
Unit SizeEach
Contents & Storage
pMIR-REPORT™ Luciferase and pMIR-REPORT™ β-gal Control Plasmid are both supplied as glycerol stocks in E. coli and should be stored at –70°C.