THE RNA Storage Solution, 10 x 1 mL - FAQs
THE RNA Storage Solution, 10 x 1 mL - FAQs
View additional product information for THE RNA Storage Solution - FAQs (AM7000, AM7001)
2 product FAQs found
Can I use RNA Storage Solution (Cat. No. AM7000, AM7001) for resuspending dye-labeled RNA?
We have not tested RNA Storage Solution with dye-labeled RNA and therefore do not recommend using it for this application.
What is the concentration of RNA Storage Solution (Cat. No. AM7000, AM7001)?
The concentration of RNA Storage Solution is 1 mM sodium citrate, pH 6.4.