Lambda Hind III dsDNA Markers
Invitrogen™

Lambda Hind III dsDNA Markers

Ambion® Lambda DNA is digested to completion with Hind III. This generates fragments of the sizes 23130, 9416, 6557, 4361,Read more
Catalog NumberQuantity
AM7720200 ug
Catalog number AM7720
Price (GBP)
296.00
Each
Quantity:
200 ug
Price (GBP)
296.00
Each
Ambion® Lambda DNA is digested to completion with Hind III. This generates fragments of the sizes 23130, 9416, 6557, 4361, 2322, 2027, 564, and 125 bp. These DNA molecular mass markers are supplied with a protocol describing easy labeling of the markers with radioisotopes. They can also be stained with ethidium bromide. One tube containing 200 μg is provided.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Gel CompatibilityAgarose Gels
Product TypeDNA Marker
Quantity200 ug
Shipping ConditionRoom Temperature
Size Range125 to 23.1 kb
Concentration0.5 mg/mL
Product LineAmbion
Unit SizeEach
Contents & Storage
Store at –20°C.

Frequently asked questions (FAQs)

How can I quantitate DNA on an agarose gel?

For fragments between 100 bp and 2,000 bp, use the Low DNA Mass Ladder. 4 µL of the Low DNA Mass Ladder corresponds to 10, 20, 40, 80, 120, and 200 ng respectively of fragments, 100, 200, 400, 800, 1,200 and 2,000 bp.

For fragments between 1,000 bp and 10,000 bp use the High DNA Mass Ladder. 4 µL of the High DNA Mass Ladder corresponds to 20, 40, 60, 80, 120, 200 ng respectively of fragments 1,000, 2,000, 3,000, 4,000, 6,000, and 10,000 bp.

To calculate the amount of DNA in 1 µg of the Lambda/Hind III standard:
(size of band in bp/48503 bp) X 1 µg = µg of DNA in band A

For example, to calculate the amount of DNA in the 564 bp band of the Lambda/Hind III standard:
(564/48,503) X 1 µg = 0.011 µg or 11 ng of DNA in the 564 bp band.