The same high-quality deoxynucleotide triphosphate solutions provided in the Ambion® RETROscript® First Strand cDNA Synthesis Kit for RT-PCR (SKU# AM1710).Read more
Have Questions?
Catalog Number
Quantity
AM8228G
300 μL
Catalog number AM8228G
Price (USD)
197.65
Online exclusive
206.00
Save 8.35
Each
Quantity:
300 μL
Price (USD)
197.65
Online exclusive
206.00
Save 8.35
Each
The same high-quality deoxynucleotide triphosphate solutions provided in the Ambion® RETROscript® First Strand cDNA Synthesis Kit for RT-PCR (SKU# AM1710). They are subject to rigorous nuclease testing as well as a functional assay to ensure high levels of incorporation. Supplied in one tube containing 0.3 mL, premixed at a concentration of 2.5 mM each.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Reverse Transcription
Label or DyeUnlabeled
Product LineAmbion
Quantity300 μL
Shipping ConditionDry Ice
Concentration2.5 mM
Unit SizeEach
Contents & Storage
Store at –20°C.
Frequently asked questions (FAQs)
Why do some dNTP mixes contain dTTP and others TTP?
Thymine is normally found as a deoxyribose. Thus, TTP and dTTP are essentially the same thing. Both types of mixes can be used in PCR reactions