Why do some dNTP mixes contain dTTP and others TTP?
Thymine is normally found as a deoxyribose. Thus, TTP and dTTP are essentially the same thing. Both types of mixes can be used in PCR reactions
Search
View additional product information for dNTP Mix (2.5 mM) - FAQs (AM8228G)
1 product FAQs found