MagMAX™ Wash Solution 2 Concentrate
Invitrogen™
MagMAX™ Wash Solution 2 Concentrate
Invitrogen MagMAX Wash Solution 2 Concentrate is a component of all the MagMAX Isolation Kits that may be purchased separately.
Have Questions?
Catalog NumberQuantity
AM864040 mL
Catalog number AM8640
Price (USD)
166.65
Online exclusive
177.00
Save 10.35
Each
Quantity:
40 mL
Price (USD)
166.65
Online exclusive
177.00
Save 10.35
Each
Invitrogen™ MagMAX™ Wash Solution 2 Concentrate is a component of all MagMAX Isolation Kits and may be purchased separately. Supplied as 40 mL of 5X concentrate. Add 160 mL of ethanol prior to use to prepare 200 mL of working Wash Solution 2.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferWash Buffer
Product LineAmbion, MagMAX
Quantity40 mL
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store MagMAX™ Wash Solution 2 Concentrate at 4°C or room temperature.
Frequently asked questions (FAQs)
How should I prepare MagMAX wash solutions 1 and 2 from standalone Wash Solution 1 Concentrate (Cat. No. AM8504) and Wash Solution 2 Concentrate (Cat. No. AM8640)?
For MagMAX Wash Solution 1 Concentrate (Cat. No. AM8504), add 70 mL isopropanol to 135 mL of wash buffer concentrate. This can be added directly to the AM8504 bottle for a total volume of 205 mL of wash solution 1. For MagMAX Wash Solution 2 Concentrate (Cat. No. AM8640), add 160 mL ethanol to 40 mL of wash buffer concentrate. This can be added directly to the AM8640 bottle for a total volume of 200 mL of wash solution 2.