RNA Fragmentation Reagents - FAQs
RNA Fragmentation Reagents - FAQs
View additional product information for RNA Fragmentation Reagents - FAQs (AM8740)
1 product FAQs found
What kind of 5' and 3' termini are left on the RNA fragments when using RNA Fragmentation Reagents (Cat. No. AM8740)?
RNA Fragmentation Reagents are designed based on RNA hydrolysis. Therefore, the resulting fragments would have 3' Phospho group and 5' OH group.