Ammonium Acetate (5 M), RNase-free
Invitrogen™

Ammonium Acetate (5 M), RNase-free

Invitrogen Molecular biology grade, 5 M Ammonium Acetate solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM9071500 mL
AM9070G100 mL
Catalog number AM9071
Price (USD)
144.65
Online exclusive
152.00
Save 7.35
Each
Quantity:
500 mL
Price (USD)
144.65
Online exclusive
152.00
Save 7.35
Each

Invitrogen Molecular biology grade, 5 M Ammonium Acetate solution is supplied in one bottle, available in multiple sizes. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Chemical Name or MaterialAmmonium Acetate
Packaging TypeBottle
Product LineAmbion
PurityMolecular Biology Grade
Quantity500 mL
Shipping ConditionRoom Temperature
Concentration5 M
FormLiquid
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

What salts can be used to precipitate total RNA? What are the advantages of each? Do they require ethanol?

The three salts that can be used are:

- Guanidine thiocyanate, which requires ethanol. Guanidine thiocyanate is a common agent used for isolating RNA and we recommend it especially for tissues high in ribonuclease activity, such as the pancreas or spleen.
- Ammonium acetate, which requires ethanol. Ammonium acetate is useful when reducing coprecipitation of unwanted dNTPs and oligosaccharides. However, it should not be used when the nucleic acid will be phosphorylated using T4 polynucleotide kinase, since this enzyme is inhibited by ammonium ions.
- Lithium chloride, which does not require ethanol. LiCl is very effective in precipitating RNA but does not efficiently precipitate protein or DNA. It also does not precipitate unincorporated nucleotides.

How can I concentrate DNA solution using ethanol precipitation?

Ethanol precipitation is frequently used for concentration of DNA solutions and for removal of protein, salt, and unincorporated nucleotides. The two most common protocols use either 0.3 M sodium acetate (0.1 volume of 3 M) or 2.5 M ammonium acetate (0.5 volume of 7.5 M), along with 2 to 2.5 volumes of ethanol. Studies at Thermo Fisher Scientific (1, 2) have shown these two salts to be equally effective for recovery of small amounts of DNA from small volumes and for removal of unincorporated nucleotides from labeling reactions.

DNA was found to precipitate readily with room temperature ethanol and room temperature centrifugation. For DNA concentrations >0.1 µg/mL, no incubation period is required. For improved recovery of DNA from dilute solutions (10 ng/mL), overnight incubation in the ethanol and extended (30 min) centrifugation is recommended. Addition of ammonium acetate to 2.5 M (without ethanol) has also been shown to be effective in precipitating proteins while leaving the DNA in solution (2).

1. Zeugin, J.A. and Hartley, J.L. (1985) FOCUS 7:4, 1.

2. Crouse, J. and Amorese, D. (1987) FOCUS 9:2, 3.