SDS, 20% Solution, RNase-free
Invitrogen™

SDS, 20% Solution, RNase-free

Ambion® molecular biology grade, SDS, 20% Solution is supplied in one bottle containing 250 mL. The buffer is certified RNase-free,Read more
Have Questions?
Catalog NumberQuantity
AM9820250 mL
Catalog number AM9820
Price (USD)
82.65
Online exclusive
86.75
Save 4.10
Each
Quantity:
250 mL
Price (USD)
82.65
Online exclusive
86.75
Save 4.10
Each
Ambion® molecular biology grade, SDS, 20% Solution is supplied in one bottle containing 250 mL. The buffer is certified RNase-free, economical, and ready-to-use. SDS is used as a component of both hybridization and wash buffers for northern and Southern blots and has many other uses around the lab.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Nucleic Acid Gel Electrophoresis & Blotting
Product LineAmbion
Product TypeSDS
PurityMolecular Biology Grade
Quantity250 mL
Shipping ConditionRoom Temperature
FormLiquid
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

What does SDS Solution (Cat. No. AM9820, AM9822, AM9823) contain?

Ambion molecular biology grade SDS Solution (Cat. No. AM9820, AM9822, AM9823) consists of Sodium Dodecyl Sulfate (SDS) and water. The solution is certified to be RNase-free and ready-to-use. AM9820: 20% SDS Solution, RNase-free, 250 mL
AM9822: 10% SDS Solution, RNase-free, 500 mL
AM9823: 10% SDS Solution, RNase-free, 1 L