Countess™ 3 FL Automated Cell Counter, 1 Ea. - FAQs
Countess™ 3 FL Automated Cell Counter, 1 Ea. - FAQs
View additional product information for Countess™ 3 FL Automated Cell Counter - FAQs (A49866, A50303, AMQAF2000, A49892, AMQAF2001, A50299, A49893)
4 product FAQs found
If we have more than one Countess at our facility, do I need to purchase a Countess 3 21 CFR Part 11 Software Module (Cat. No A51025) for each Countess 3/3FL Automated Cell Counter?
Yes. Each Countess 3 21 CFR Part 11 Software Module (Cat. No. A51025) purchase covers only one Countess instrument.
What is the activation key for the Countess 3/3 FL software?
The activation key is a file that can be downloaded from the Countess Software Downloads web page upon entering the Order ID and the instrument serial number on the prompt screen.
I ordered the Countess 3 21 CFR Part 11 Software Module (Cat. No. A51025), what will I receive with my purchase?
With your purchase of the Countess 3 21 CFR Part 11 Software Module (Cat. No. A51025) you will receive an envelope with instructions on how to create a license key via Thermo Fisher Cloud and how to install this on your Countess 3 or 3 FL Automated Cell Counter.
Can I use crystal violet with the Countess 3 (Cat. No. AMQAX2000) and 3 FL Automated Cell Counter (Cat. No. AMQAF2000)?
Yes. Crystal violet is compatible with Countess 3 (Cat. No. AMQAX2000) and 3 FL Automated Cell Counter (AMQAF2000) under brightfield mode detection.
Find additional tips, troubleshooting help, and resources within our Cell Viability, Proliferation, Cryopreservation, and Apoptosis Support Center.