VetMAX™ Avian Metapneumovirus Real-Time PCR Reagents
Thermo Scientific™

VetMAX™ Avian Metapneumovirus Real-Time PCR Reagents

Real-time PCR is a widely used molecular biology technique for the amplification of nucleic acid targets in animal-derived samples.
Have Questions?
Catalog NumberKit Type
APVP50Avian Metapneumovirus Real Time PCR Kit
Catalog number APVP50
Price (USD)
-
Kit Type:
Avian Metapneumovirus Real Time PCR Kit

Real-time PCR is a widely used molecular biology technique for the amplification of nucleic acid targets in animal-derived samples.

This reagent set includes:

  • Two reagent mixes containing oligonucleotides targeting APV A/B and APV C sequences, along with an internal process control (IPC).
  • One external reference material containing APV A/B/C target sequences of the same nature as the target material (inactivated bacteria or virus) for use in workflow verification and assay performance assessment.
  • One exogenous internal process control to be added during nucleic acid extraction for monitoring extraction workflow consistency.
  • One quality control certificate.
Specifications
Kit TypeAvian Metapneumovirus Real Time PCR Kit
No. of Tests50 Reactions
Product LineVetMAX
Product TypeReal-Time PCR Reagent
Quantity50 Reactions
Sample TypeLung, Tracheal Swab
SpeciesPoultry
TargetG/SH
Target Organism ClassAvian Metapneumovirus
Detection MethodPrimer-probe
For Use With (Application)Real Time PCR (qPCR), Animal Health
GC-Rich PCR PerformanceLow
Unit SizeEach