DL-Asparagine monohydrate, 98%
Thermo Scientific Chemicals

DL-Asparagine monohydrate, 98%

CAS: 69833-18-7 | C4H8N2O3 | 132.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B20273.36
also known as B20273-36
Price (USD)
303.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
303.00
Each
Chemical Identifiers
CAS69833-18-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
DL-Asparagine monohydrate is used as a pharmaceutical intermediate. It is used to prepare DL-asparaginium nitrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Asparagine monohydrate is used as a pharmaceutical intermediate. It is used to prepare DL-asparaginium nitrate.

Solubility
Soluble in water, acids, and alkalis.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Kaoru Harada. The Optical Resolution of DL-Aspartic Acid, DL-Glutamic Acid, DL-Asparagine and DL-Glutamine by Preferential Crystallization. Bulletin of the Chemical Society of Japan. 1965, 38 (9), 1552-1555.
  2. N. Moussa Slimane; A. Cherouana; L. Bendjeddou; S. Dahaoui and C. Lecomte. DL-Asparaginium nitrate. Acta Cryst. 2009, E65, o2180-o2181.