6-Chloro-3-indolyl-beta-D-galactopyranoside, 98%
6-Chloro-3-indolyl-beta-D-galactopyranoside, 98%
6-Chloro-3-indolyl-beta-D-galactopyranoside, 98%
Thermo Scientific Chemicals

6-Chloro-3-indolyl-beta-D-galactopyranoside, 98%

CAS: 138182-21-5 | C14H16ClNO6 | 329.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.1 g
0.5 g
Catalog number B21131.02
also known as B21131-02
Price (USD)
310.65
Online Exclusive
345.00
Save 34.35 (10%)
Each
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
310.65
Online Exclusive
345.00
Save 34.35 (10%)
Each
Chemical Identifiers
CAS138182-21-5
IUPAC Name(2S,3R,4S,5R,6R)-2-[(6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC14H16ClNO6
InChI KeyOQWBAXBVBGNSPW-MBJXGIAVSA-N
SMILESOC[C@H]1O[C@@H](OC2=CNC3=CC(Cl)=CC=C23)[C@H](O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Optical Rotation-43.5 ± 2.0? (C=1 in methanol)
Assay (HPLC)≥97.5%
Used as a pharmaceutical intermediate to reduce toxicity and as an enzyme substrate in diagnostic reagents. Salmon-gal is used in conjunction with IPTG for detection of β-galactosidase activity in bacterial colonies in a colometric assay to detect recombinants (white) from the non-recombinants (SALMON). In conjunction with X-glu, salmon-gal is useful for simultaneous detection of GUS and Lac activities on the same plate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a pharmaceutical intermediate to reduce toxicity and as an enzyme substrate in diagnostic reagents. Salmon-gal is used in conjunction with IPTG for detection of β-galactosidase activity in bacterial colonies in a colometric assay to detect recombinants (white) from the non-recombinants (SALMON). In conjunction with X-glu, salmon-gal is useful for simultaneous detection of GUS and Lac activities on the same plate.

Solubility
Insoluble in water. Soluble in DMSO and DMFO, methanol.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. Simonson MS, et al. Rapid characterization of growth-arrest genes in transient transfection assays.BioTechniques.199518(3), 434-6, 438, 440-2.
  2. Aimin Liu, et al. Alteration of limb and brain patterning in early mouse embryos by ultrasound-guided injection of Shh-expressing cells.Mechanisms of Development.199875(1-2), 107-115.