N-Acetyl-DL-methionine, 99%
N-Acetyl-DL-methionine, 99%
N-Acetyl-DL-methionine, 99%
Thermo Scientific Chemicals

N-Acetyl-DL-methionine, 99%

CAS: 1115-47-5 | C7H13NO3S | 191.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number B21866.0B
also known as B21866-0B
Price (USD)
92.65
Online exclusive
103.00
Save 10.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
92.65
Online exclusive
103.00
Save 10.35
Each
Chemical Identifiers
CAS1115-47-5
IUPAC Name2-acetamido-4-(methylsulfanyl)butanoic acid
Molecular FormulaC7H13NO3S
InChI KeyXUYPXLNMDZIRQH-UHFFFAOYNA-N
SMILESCSCCC(NC(C)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Appearance (Color)White
Melting Point (clear melt)112.0-118.0?C
N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.

Solubility
Soluble in water, ethanol, ethyl acetate.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. S. Kominami; K. Akasaka; H. Umegaki; H. Hatano. Photo-induced reversible hole-transfer in a γ-irradiated single crystal of N-acetyl DL-methionine: Existence of two hole traps in a molecule. Chemical Physics Letters. 1971, 9 (6), 510-513.
  2. H. E. Amos; C. O. Little; G. A. Digenis; G. T. Schelling; R. E. Tucker and G. E. Mitchell. Methionine, DL-Homocysteine Thiolactone and N-Acetyl-DL-Methionine for Ruminants. Journal of Animal Science. 1943, 39 (3), 612-617.