Mucic acid, 98%
Mucic acid, 98%
Mucic acid, 98%
Thermo Scientific Chemicals

Mucic acid, 98%

CAS: 526-99-8 | C6H10O8 | 210.138 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B24170.36
also known as B24170-36
Price (USD)
209.65
Online Exclusive
233.00
Save 23.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
209.65
Online Exclusive
233.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS526-99-8
IUPAC Name2,3,4,5-tetrahydroxyhexanedioic acid
Molecular FormulaC6H10O8
InChI KeyDSLZVSRJTYRBFB-UHFFFAOYNA-N
SMILESOC(C(O)C(O)C(O)=O)C(O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Sulfated Ash≤0.1%
Appearance (Color)White to pale cream
Assay from Supplier's CofAConforms to structure
Solution Test4% wt (1N NaOH solution): Clear, colorless solution
View more
Mucic acid is used to replace tartaric acid in self-rising flour or fizzies. It is also used as a precursor of adipic acid en route to nylon by a rhenium-catalyzed deoxydehydration reaction. It replaces potassium bitartrate in baking powder and also used for manufacture of granular effervescing salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mucic acid is used to replace tartaric acid in self-rising flour or fizzies. It is also used as a precursor of adipic acid en route to nylon by a rhenium-catalyzed deoxydehydration reaction. It replaces potassium bitartrate in baking powder and also used for manufacture of granular effervescing salts.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. George A. Jeffrey.; Richard A. Wood. The crystal structure of galactaric acid (mucic acid) at -147°: An unusually dense, hydrogen-bonded structure. Carbohydrate Research. 1982, 108 (2), 205-211.
  2. G. Avigad.; C. Asensio.; D. Amaral.; B.L. Horecker. Galacto-dialdose production with an enzyme from the mold Polyporuscircinatus. Biochemical and Biophysical Research Communications. 1961, 4 (6), 474-477.