N-Boc-L-phenylalaninol, 99%
N-Boc-L-phenylalaninol, 99%
N-Boc-L-phenylalaninol, 99%
Thermo Scientific Chemicals

N-Boc-L-phenylalaninol, 99%

CAS: 66605-57-0 | C14H21NO3 | 251.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B25019.03
also known as B25019-03
Price (USD)
22.65
Special offer
Online exclusive
Ends: 30-Jun-2026
26.30
Save 3.65 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
22.65
Special offer
Online exclusive
Ends: 30-Jun-2026
26.30
Save 3.65 (14%)
Each
Chemical Identifiers
CAS66605-57-0
IUPAC Nametert-butyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
Molecular FormulaC14H21NO3
InChI KeyLDKDMDVMMCXTMO-LBPRGKRZSA-N
SMILESCC(C)(C)OC(=O)N[C@H](CO)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.5%
Melting Point (clear melt)92.0-98.0?C
Optical Rotation-27.0 ± 3.0? (C=1 in methanol)
N-Boc-L-phenylalaninol is employed in the synthesis of biochemically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-phenylalaninol is employed in the synthesis of biochemically active compounds.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Nicholas J. Miles; Peter G. Sammes; Peter D. Kennewell and Robert Westwood . On the double bond isostere of the peptide bond: preparation of modified di- and tri-peptides incorporating proline and alanine analogues. J. Chem. Soc., Perkin Trans. 1. 1985, 2299-2305.
  2. Dario Neri Prof. Dr.; Giacomo Fossati and Matteo Zanda Dr. Efforts toward the Total Synthesis of Tubulysins: New Hopes for a More Effective Targeted Drug Delivery to Tumors. ChemMedChem. 2006, 1 (2), 175-180.