BrdU Monoclonal Antibody (MoBU-1) - FAQs
BrdU Monoclonal Antibody (MoBU-1) - FAQs
View additional product information for BrdU Monoclonal Antibody (MoBU-1) - FAQs (B35141)
1 product FAQs found
Does BrdU Mouse Monoclonal Antibody (Clone MoBU-1) cross-react with IdU?
We do not have any information regarding the cross-reactivity of the BrdU Mouse Monoclonal Antibody (Clone MoBU-1) with IdU.