Reaction Buffer for phi29 DNA Polymerase (10X)
Thermo Scientific™

Reaction Buffer for phi29 DNA Polymerase (10X)

Thermo Scientific™ 10X Buffer for phi29 DNA Polymerase is the optimal buffer recommended for use with highly processive phi29 DNA polymerase.
Have Questions?
Catalog NumberQuantity
B625 x 1 mL
Catalog number B62
Price (USD)
22.65
Online exclusive
23.35
Each
Quantity:
5 x 1 mL
Request bulk or custom format
Price (USD)
22.65
Online exclusive
23.35
Each

Thermo Scientific™ 10X Buffer for phi29 DNA Polymerase is the optimal buffer recommended for use with highly processive phi29 DNA polymerase.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Concentration10X
Shipping ConditionDry Ice
For Use With (Application)Isothermal Amplification
Product TypeReaction Buffer
Quantity5 x 1 mL
Research CategoryTraditional Cloning
Unit SizeEach
Contents & Storage

• 10X Reaction buffer for Phi29 DNA Polymerase (5 x 1.0 mL)

Store at –5 to –30°C.