본 제품은 액체 질소로 배송 및 보관되는 세포 관련 제품으로, 해당 제품 구매 시 세포 사용 가이드라인을 반드시 확인해주십시오. 세포사용가이드라인 다운로드
Product Image
Gibco™

Sf9 Cells in Grace's

Gibco™ Sf9 cells are commonly used to isolate and propagate recombinant baculoviral stocks and to produce recombinant proteins. The cells자세히 알아보기
Have Questions?
카탈로그 번호수량
B825011 mL
카탈로그 번호 B82501
제품 가격(KRW)
617,000
온라인 행사
Ends: 30-Jun-2026
705,000
할인액 88,000 (12%)
Each
수량:
1 mL
제품 가격(KRW)
617,000
온라인 행사
Ends: 30-Jun-2026
705,000
할인액 88,000 (12%)
Each
Gibco™ Sf9 cells are commonly used to isolate and propagate recombinant baculoviral stocks and to produce recombinant proteins. The cells originated at the USDA Insect Pathology Laboratory, from the parental IPLBSF-21 (Sf21) cell line, which was derived from the pupal ovarian tissue of the fall army worm, Spodoptera frugiperda. Gibco™ Sf9 cells (frozen in Grace's Insect Medium) feature:
• Recombinant protein expression from a variety of expression systems
• Good growth in adherent or suspension culture
• Small, regular size that generates even monolayers and plaques
• Quality and performance testing

Recombinant protein expression from a variety of expression systems
High levels of protein expression in Sf9 cells can be obtained using either the BaculoDirect™ Baculovirus Expression System, the Bac-to-Bac™ Baculovirus Expression System, or the InsectDirect™ Expression System.

Good growth in adherent or suspension culture
Protocols for adherent or suspension growth in Gibco™ Supplemented Grace's Insect Medium are available in the product manual. Cultures can easily be transferred between the two conditions for ease of workflow.

Small, regular size that generates even monolayers and plaques
Gibco™ Sf9 cells generate a clean, even monolayer and plaques due to their small, round, regular size. Other cells often form more irregular monolayers and plaques.

Quality and performance testing
Each lot of Gibco™ Sf9 cells is tested for cell growth and viability post-recovery from cryopreservation. In addition, the Master Seed Bank has been tested for contamination of bacteria, yeast, mycoplasma and virus and has been characterized by isozyme and karyotype analysis.

Caution: Handle as potentially biohazardous material under at least Biosafety Level 2 containment. This product contains Dimethyl Sulfoxide (DMSO), a hazardous material. Review the Material Safety Data Sheet before handling.
For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
사양
제품 유형Insect Cells
수량1 mL
세포주Sf9
셀 유형Insect Cells
S. frugiperda
Unit SizeEach
구성 및 보관
Storage conditions: Liquid nitrogen (vapor phase)
Shipping conditions: Frozen

자주 묻는 질문(FAQ)

What is the procedure to thaw frozen insect cells?

The following protocol describes a general procedure for thawing cryopreserved cells. For detailed protocols, always refer to the cell-specific product insert.

1. Remove the cryovial containing the frozen cells from liquid nitrogen storage and immediately place it into a 37°C water bath.
2. Quickly thaw the cells (< 1 minute) by gently swirling the vial in the 37°C water bath until there is just a small bit of ice left in the vial.
3. Transfer the vial into a laminar flow hood. Before opening, wipe the outside of the vial with 70% ethanol.
4. Transfer the desired amount of pre-warmed complete growth medium appropriate for your cell line dropwise into the centrifuge tube containing the thawed cells.
5. Centrifuge the cell suspension at approximately 200 x g for 5-10 minutes. The actual centrifugation speed and duration varies depending on the cell type.
6. After the centrifugation, check the clarity of supernatant and visibility of a complete pellet. Aseptically decant the supernatant without disturbing the cell pellet.
7. Gently resuspend the cells in complete growth medium, and transfer them into the appropriate culture vessel and into the recommended culture environment.

Note: The appropriate flask size depends on the number of cells frozen in the cryovial, and the culture environment varies based on the cell and media type.

Why does the Insect cell line manual state: "Cells should be maintained at 27 degrees C in a non-humidified environment."

Insect cells do not require CO2 or high humidity to grow, they can grow in a lab drawer at room temperature. We recommend this so people don't waste CO2 and other resources necessary for maintaining a tissue culture incubator. It should be noted, however, that the cells will grow in a humidified incubator.

When growing Sf9 cells in a bioreactor, can I use a glass vessel that has been cleaned and autoclaved and then reused or do I need to use a disposable vessel?

Yes, you can grow Sf9 cells in glass vessels. The only concern would be if your glass vessels are not clean enough and there may be residual detergent left which will hurt your cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are spindle-shaped Sf9 cells bad? How do I get rid of them?

These cells appear after cultures have been grown for several weeks. These do not seem to be detrimental to plating of high titer stocks or expression. However, if they are in high numbers, it may indicate that the cells are becoming old and that the culture should be re-started with a new stock of frozen cells.

What are small particles that appear after a few days when growing Sf9 cells? I am not using Gibco Fungizone reagent, but I am using gentamicin. Are they yeast?

Yellow particles could be cell organelles, aggregates, or debris. We see this when we first thaw frozen cells. To avoid this, you can let the shaker culture sit for 5 minutes, then transfer top 1/3 to a new flask, making sure to count cells first. You can also use heparin at up to 200 U/mL to decrease aggregation. Pluronic solution at a final concentration of 0.2% can also be used to decrease shearing, and increase shake speed to 100-120 rpm. Recently thawed cells seem to be breaking up and releasing small vesicles, as observed under high magnification. To reduce the amount of those small particles, cells need to be rapidly but completely defrosted for successful thawing to take place. Also:

1. Place vial on ice during transfer from water bath to sterile hood.
2. Pipette as gently as possible because cells shear easily due to larger surface area.
3. Cells may not have been placed in cold media after removal from defrosted vial into flask.
4. Media may not have been changed after 30-45 minutes once a majority of cells had attached. Media change should be with pre-warmed media (27degrees C). 10% DMSO in freezing medium will kill the cells if left on them for long periods of time (1 hour seems to be a maximum).
5. Lastly, cells should be checked for contamination. To do so, plate a small portion of culture in a T-25 flask and incubate for 3 days, checking for cloudiness.

인용 및 참조 문헌 (47)

인용 및 참조 문헌
Abstract
OX40 stimulation by gp34/OX40 ligand enhances productive human immunodeficiency virus type 1 infection.
Authors: Takahashi Y; Tanaka Y; Yamashita A; Koyanagi Y; Nakamura M; Yamamoto N;
Journal:J Virol
PubMed ID:11435553
'OX40 is a member of the tumor necrosis factor (TNF) receptor superfamily and known to be an important costimulatory molecule expressed on activated T cells. To investigate the role of costimulation of OX40 in human immunodeficiency virus type 1 (HIV-1) infection by its natural ligand, gp34, the OX40-transfected ACH-2 cell ... More
Cytoskeletal changes regulated by the PAK4 serine/threonine kinase are mediated by LIM kinase 1 and cofilin.
Authors: Dan C; Kelly A; Bernard O; Minden A;
Journal:J Biol Chem
PubMed ID:11413130
'PAK4 is the most recently identified member of the PAK family of serine/threonine kinases. PAK4 differs from other members of the PAK family in sequence and in many of its functions. Previously, we have shown that an important function of this kinase is to mediate the induction of filopodia in ... More
The enzymological basis for resistance of herpesvirus DNA polymerase mutants to acyclovir: relationship to the structure of alpha-like DNA polymerases.
Authors:Huang L, Ishii KK, Zuccola H, Gehring AM, Hwang CB, Hogle J, Coen DM
Journal:Proc Natl Acad Sci U S A
PubMed ID:9892653
'Acyclovir (ACV), like many antiviral drugs, is a nucleoside analog. In vitro, ACV triphosphate inhibits herpesvirus DNA polymerase by means of binding, incorporation into primer/template, and dead-end complex formation in the presence of the next deoxynucleoside triphosphate. However, it is not known whether this mechanism operates in vivo. To address ... More
Expression and characterization of functionally active recombinant perforin produced in insect cells.
Authors:Liu CC, Persechini PM, Young JD
Journal:J Immunol
PubMed ID:8617952
'A key cytolytic mediator used by killer lymphocytes, perforin (also known as pore-forming protein or cytolysin), has been shown to be capable of undergoing polymerization to form pores in cell membranes and cause osmotic lysis of target cells. Although perforin has been purified from killer lymphocytes and the coding gene ... More
NS5A, a nonstructural protein of hepatitis C virus, binds growth factor receptor-bound protein 2 adaptor protein in a Src homology 3 domain/ligand-dependent manner and perturbs mitogenic signaling.
Authors:Tan SL, Nakao H, He Y, Vijaysri S, Neddermann P, Jacobs BL, Mayer BJ, Katze MG
Journal:Proc Natl Acad Sci U S A
PubMed ID:10318918
'Although hepatitis C virus (HCV) infection is an emerging global epidemic causing severe liver disorders, the molecular mechanisms of HCV pathogenesis remain elusive. The NS5A nonstructural protein of HCV contains several proline-rich sequences consistent with Src homology (SH) 3-binding sites found in cellular signaling molecules. Here, we demonstrate that NS5A ... More