eBioscience™ Propidium Iodide - FAQs
eBioscience™ Propidium Iodide - FAQs
View additional product information for eBioscience™ Propidium Iodide - FAQs (BMS500PI)
1 product FAQs found
What is the concentration of eBioscience Propidium Iodide?
eBioscience Propidium Iodide is supplied at a concentration of 0.02 mg/mL and we recommend using 10 µL in a total staining volume of 200 µL.