Nebulizers for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Thermo Scientific™
Nebulizers for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Select a Thermo Scientific™ Nebulizer to meet your laboratory needs from this range compatible with Element HR-ICP-MS and iCAP MX and iCAP Q/Qnova ICP-MS systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
BRE0063796PFA C50 Nebulizer
1162030PFA LC Nebulizer
1161990Backflush Kit
1317090PFA-ST MicroFlow Nebulizer
BRE0009386MicroMist Nebulizer 0.4 mL/min
1600165PFA LF Nebulizer, 50 μL/min
1600342PFA LF Nebulizer 100 μL/min
1600427PFA LF Nebulizer, 20 μL/min
4600356Burgener Mira Mist Nebulizer
4600495Burgener Ari Mist Nebulizer
1161870MicroMist Nebulizer 0.2 mL/min
BRE0061133PFA Nebulizer C400d
BRE0063797PFA C100 Nebulizer
BRE0063798PFA C250 Nebulizer
BRE0063792Uptake Line C400d Nebulizer
Catalog number BRE0063796
Price (USD)
3,490.00
Each
Includes:
PFA C50 Nebulizer
Price (USD)
3,490.00
Each
We offer a range of nebulizers compatible with the Thermo Scientific™ Element, iCAP MX and iCAP Q/Qnova Series ICP-MS systems to meet the needs of your laboratory. The standard cyclonic spray chamber accepts all types of nebulizers with a 6 mm outside diameter.
Optimize data quality and minimize recalibrations with high-performance nebulizer options
  • Choose glass nebulizers for standard applications
  • Choose PFA nebulizers for ultra-trace or highly concentrated acids and samples containing HF
  • Choose low-flow nebulizers for organic sample analysis and cold plasma applications
  • Choose high-solids nebulizers to minimize salt build-up and nebulizer clogging for highly concentrated samples
Specifications
For Use With (Equipment)iCAP MX Series ICP-MS Instruments
IncludesPFA C50 Nebulizer
MaterialPFA
TypeNebulizer
Unit SizeEach