Sample Handling Kits for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Thermo Scientific™
Sample Handling Kits for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Get accurate ICP-MS results using dedicated sample handling kits for the Thermo Scientific™ Element™ Series HR-ICP-MS and Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
BRE0064559iCAP MX Acid Resistance KitiCAP MX Series ICP-MS
1067531Organic Matrix AccessoryElement Series HR-ICP-MS
1159440Matrix KitElement Series HR-ICP-MS
1162170Acid-Resistant Sample IntroElement Series HR-ICP-MS
1324711Organic Matrix KitiCAP MX and iCAP Q/Qnova
1324720Acid Resistant Kit Q/QnovaiCAP Q/Qnova ICP-MS
Catalog number BRE0064559
Price (USD)
10,300.00
Each
Type:
iCAP MX Acid Resistance Kit
For Use With (Equipment):
iCAP MX Series ICP-MS
Price (USD)
10,300.00
Each
Enhance your analysis with dedicated Thermo Scientific™ Sample Handling Kits for the Element Series HR-ICP-MS, the iCAP MX and iCAP Q/Qnova Series ICP-MS instruments.
Enable the routine analysis of organic solvents, aggressively acidic or highly complex sample matrices with these convenient, complete sample handling accessory kits.
Enhance System Performance with Tailored Sampling Kits
Organic Matrix Accessory Kit for Element Series HR-ICP-MS
Organic solvents are easily handled with this kit comprising:
  • Mass flow controller for use with oxygen
  • Dual spray chamber
  • Skimmer and sample cones (platinum tipped) not included
  • Nebulizer and injector
  • Tubing and connector
Acid-Resistant Sample Introduction System
For the routine analysis of aggressive sample matrices, this kit includes:
  • Mass flow controller for use with argon gas
  • Special pumping system with synthetic oil
  • Sampler and skimmer cones (platinum tipped) included
Complex Matrix Sample Handling Kit for Element Series HR-ICP-MS
Enable the routine analysis of highly complex matrices with this product bundle containing:
  • MicroMist nebulizer
  • Twinnabar cyclonic spray chamber
  • Injectors 2.2 mm
  • Tubing and connectors
Specifications
For Use With (Equipment)iCAP MX Series ICP-MS
TypeiCAP MX Acid Resistance Kit
Unit SizeEach