iCAP™ PRO ICP-OES Radial - FAQs
iCAP™ PRO ICP-OES Radial - FAQs
View additional product information for iCAP™ PRO ICP-OES and iCAP™ PRO X ICP-OES - FAQs (BRE731403, BRE731402, BRE731401, BRE731400)
12 product FAQs found
What are the main differences between the iCAP PRO, iCAP PRO X, iCAP PRO XP, and iCAP PRO XPS models?
The main differences among the Thermo Scientific iCAP PRO models are based on their specific features and intended applications:
  • iCAP PRO: For routine laboratory analysis with fast start-up, easy-to-use software, and robust performance. It simplifies analysis in laboratories performing standardized analysis for a low number of samples.
  • iCAP PRO X: For high-matrix trace elemental analysis with sensitive multi-element detection, ideal for challenging samples. It helps laboratories with moderate throughput and established methods to improve performance and reliability.
  • iCAP PRO XP: Flexibility for method optimization and achieves advanced performance using an enhanced ultraviolet (eUV) analysis mode, which measures the UV region of the spectrum with greater sensitivity. Fast data acquisition times allow for high throughput labs to stay competitive.
  • iCAP PRO XPS: Exceptional throughput and versatility, meeting specific regulatory requirements with high-speed analysis and excellent stability. With short integration times and high flexibility, this system is appropriate for testing labs that require the high throughput and performance.
What accessories are available for the Thermo Scientific iCAP PRO Series ICP-OES?
A wide range of accessories, including nebulizers, spray chambers, and sample introduction kits, tailored for different sample types and applications, are available. These accessories enhance the instrument's versatility and performance.
How do iCAP PRO Series ICP-OES systems support compliance with regulatory standards?
The Thermo Scientific iCAP PRO Series ICP-OES systems support full traceability of results and workflows, complying with CFR 21 Part 11. They include features like electronic signatures and audit trails to ensure regulatory compliance.
What is the Get Ready feature in the iCAP PRO Series ICP-OES?
The Get Ready process, available with Thermo Scientific Qtegra Intelligent Scientific Data Solution (ISDS) Software simplifies instrument set up by automating a series of routine tasks. After assessing results from an initial performance check, Get Ready automatically triggers any required corrective actions until the instrument is ready to perform analysis.
What types of samples can Thermo Scientific iCAP PRO Series ICP-OES systems analyze?
Thermo Scientific iCAP PRO Series ICP-OES systems can analyze a wide range of samples, including drinking water, high-matrix samples like sludge, high-purity chemicals, and samples with high total dissolved solids (TDS).
Are iCAP PRO Series ICP-OES instruments easy to use?
Yes. Thermo Scientific iCAP PRO Series ICP-OES instruments are designed with an intuitive interface, automated setup features, and minimal maintenance requirements. Routine analysis is simplified by automated method development features within the Thermo Scientific Qtegra ISDS Software, which can use plug-ins to further enhance the system capabilities.
What kind of features can I expect with theiCAP PRO Series ICP-OES system?
The Thermo Scientific iCAP PRO Series ICP-OES performs simple analysis of complex line-rich samples using a spectrometer resolution of < 7 pm at 200 nm. The system uses fewer optical surfaces to minimize reflective losses and maximize light transmission from plasma to detector for superior detection limits. Advanced automatic stabilization dramatically reduces the instrument’s warm-up time without impacting analysis times. The high-speed random access CID allows complete simultaneous access to the spectrum between 167.021 to 852.145 nm, regardless of concentration.
What is the Thermo Scientific iCAP PRO Series ICP-OES?
The Thermo Scientific iCAP PRO Series ICP-OES is a family of inductively coupled plasma-optical emission spectrometers instruments designed for robust, multi-elemental analysis. The instruments are designed to combine optimal performance with flexibility and offer high sensitivity, precision for a wide range of applications, from environmental analysis to industrial quality control.
What are the dimensions of the Thermo Scientific iCAP PRO Series ICP-OES?
W 615 mm x D 690 mm x H 933 mm; Footprint = 0.42 m2
What is the dynamic range of the Thermo Scientific iCAP PRO Series ICP-OES detector?
The CID821 detector supports a wide dynamic range of nine orders of magnitude, enabling analysis from % levels to sub-ppb concentrations.
What wavelength range does the Thermo Scientific iCAP PRO Series ICP-OES instrument cover?
The Thermo Scientific iCAP PRO Series ICP-OES covers 167.021 to 852.145 nm, enabling detection of Al at 167 nm and Na/K at their sensitive lines.
How fast is the start-up time of the Thermo Scientific iCAP PRO Series ICP-OES?
The Thermo Scientific iCAP PRO Series ICP-OES starts up in just 30 minutes from power-off and can be ready in as little as 5 minutes from standby (model dependent), helping laboratories maximize uptime.