Need a safer alternative to Trypan Blue? Try SafeCount™, a non-toxic alternative for cell counting applications that functions identically to Trypan Blue.
Countess™ Cell Counting Chamber Slides
Invitrogen™

Countess™ Cell Counting Chamber Slides

Countess Cell Counting Chamber Slides are conveniently packaged for use with the Countess Automated Cell Counters. Each disposable and reusable slide features two enclosed chambers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeIncludesQuantity
C10314Cell Counting Chamber SlideTrypan Blue Stain, 100 mL Bottle2500 Slides
A25750Reusable SlideNo Stain1 Glass Slide
A40008469Cell Counting Chamber SlideSafeCount Stain, 2 x 1 mL Vials50 Slides
C10283Cell Counting Chamber SlideNo Stain50 Slides
C10228Cell Counting Chamber SlideTrypan Blue Stain, 2 x 1 mL Vials50 Slides
C10312Cell Counting Chamber SlideTrypan Blue Stain, 20 x 1 mL Vials500 Slides
C10313Cell Counting Chamber SlideTrypan Blue Stain, 100 mL Bottle1250 Slides
C10315Cell Counting Chamber SlideTrypan Blue Stain, 2 x 100 mL Bottles5000 Slides
Catalog number C10314
Price (USD)
5,403.80
Special offer
Online exclusive
Ends: 31-Dec-2026
6,590.00
Save 1,186.20
Each
Type:
Cell Counting Chamber Slide
Includes:
Trypan Blue Stain, 100 mL Bottle
Quantity:
2500 Slides
Price (USD)
5,403.80
Special offer
Online exclusive
Ends: 31-Dec-2026
6,590.00
Save 1,186.20
Each
The Countess Chamber Slides are designed for use with the Countess 3 and 3FL automated cell counters.

The disposable Countess Cell Counting Chamber Slides offer convenience in automated cell counting for processing multiple cell samples, busy cell culture facilities, and easy disposal of contaminated or hazardous samples. They also eliminate the risk of contamination from residual samples. After use, Countess slides can be disposed of as standard biohazard waste.

The Countess Reusable Slide is a green and cost-efficient alternative to disposable slides for automated cell counting. The reusable slide enables researchers and cell culture facilities to reduce costs and generate less waste per year, making it affordable for labs to automate their cell counts. The reusable slide is cleaned and dried between counts, just as with a hemocytometer.

Each Countess Cell Counting Chamber Slide has two separate, enclosed chambers for counting one sample in duplicate or two different samples. Cell counting occurs in a central location of the counting chamber. The volume counted is 0.4 μL, the same as counting four 1-mm2 squares in a standard hemocytometer.

The Countess Automated Cell Counter works in three simple steps:

  • Mix 10 μL of sample with 10 μL of a cell viability stain or other desired reagent, including ReadyCount™ stains, and pipet into a Countess chamber slide
  • Insert the slide into the instrument
  • Press 'Count'

That's it—get the total cell count, percent viable cells, and average cell size in as little as 30 seconds.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Depth (Metric)100 μm (Chamber)
Dimensions75 x 25 x 1.8 mm (Chamber)
For Use With (Equipment)Countess Automated Cell Counter
Holds10 μL/Chamber
IncludesTrypan Blue Stain, 100 mL Bottle
Positions2 Chambers/Slide
Quantity2500 Slides
Product LineCountess
TypeCell Counting Chamber Slide
Unit SizeEach
Contents & Storage
• Countess™ Cell Counting Chamber Slides, 50 x 50 slides
• Trypan blue stain, 100 mL bottle

Each slide contains two cell counting chambers.
For use at room temperature under normal laboratory conditions.

Frequently asked questions (FAQs)

Do you offer an alternative for Countess Cell Counting Chamber Slides (Cat. no. C10283) for Countess 3 and Countess 3 FL Automated Cell Counter?

Yes. The alternative options for Countess Cell Counting Chamber Slides (Cat. No. C10283) and for Countess 3 and Countess 3 FL Automated Cell Counter can be found using the link below:

Countess Cell Counting Chamber Slides (Cat. Nos. C10228, C10312, C10313, C10314, C10315)

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What Countess Automated Cell Counter instruments are compatible with the Countess Cell Counting Chamber Slides (Cat. No. C10228, C10312, C10314, C10315)?

The Countess Cell Counting Chamber Slides (Cat. No. C10228, C10312, C10314, C10315) are compatible with all Countess Automated Cell Counter instruments.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center .