CellROX™ Reagent Variety Pack, for oxidative stress detection - FAQs
CellROX™ Reagent Variety Pack, for oxidative stress detection - FAQs
View additional product information for CellROX™ Reagent Variety Pack, for oxidative stress detection - FAQs (C10448)
1 product FAQs found
What dyes can I use to detect reactive oxygen species (ROS) in my bacteria?
Many dyes that are used on mammalian cells have also been shown to be useful in bacterial cells. For example, CellROX Deep Red Reagent has been shown to work in B. subtilis (see Reference: http://www-brs.ub.ruhr-uni-bochum.de/netahtml/HSS/Diss/RaatschenNadja/diss.pdf). If you are interested in a particular dye, but are not sure if it will work on your bacteria, literature searches are the best way to check to see if it has been tested. If not, then it may be worth testing yourself.