I am not getting any signal with the Click-iT EdU Cell Proliferation Assay for Microplates. What can cause this?
If the Amplex UltraRed reagent was spontaneously oxidized, all samples would have high levels of signal. The lack of any signal indicates that some component in the reaction is non-functional or little or no EdU was incorporated by the cells. The HRP-azide (Component B), the Click-iT EdU reaction additive (Component F), and the hydrogen peroxide (Component J) are the most labile components in the kit. HRP can be denatured and inhibited by sodium azide. The Click-iT Edu reaction additive (Component F) should be diluted only shortly before use and dilute solutions should not be stored. Hydrogen peroxide does spontaneously degrade over time, but more readily when diluted; prepare the hydrogen peroxide stock solution shortly before use and do not store.